Product Details

SNP ID
rs116606308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82389064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACCTCGTGTTTCGGCTTTGCTTT[C/T]GCAATGCACGGCTGTGCGCTCGGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105376790 PubMed Links

Gene Details

Gene
LOC105376790
Gene Name
uncharacterized LOC105376790
There are no transcripts associated with this gene.

Gene
OGFOD3
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024648.2 Intron NP_078924.1
NM_175902.4 Intron NP_787098.3

View Full Product Details