Product Details

SNP ID
rs113701085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:10523142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCACTTTTGCCTGAAGTTTATCT[A/G]CCAAATCTTGAAGCCTGAGAATATT
Phenotype
MIM: 160730 MIM: 160740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYH1 PubMed Links

Gene Details

Gene
MYH1
Gene Name
myosin heavy chain 1
There are no transcripts associated with this gene.

Gene
MYH2
Gene Name
myosin heavy chain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100112.1 5749 Missense Mutation GCA,GTA A1874V NP_001093582.1
NM_017534.5 5749 Missense Mutation GCA,GTA A1874V NP_060004.3
Gene
MYHAS
Gene Name
myosin heavy chain gene cluster antisense RNA
There are no transcripts associated with this gene.

View Full Product Details