Product Details

SNP ID
rs116911386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2036598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTGGACATCCGGATAGACACTA[C/T]ACACCTCCTGGACTCTCTCCGCCTC
Phenotype
MIM: 603527 MIM: 607896
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPH1 PubMed Links

Gene Details

Gene
DPH1
Gene Name
diphthamide biosynthesis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001383.3 503 Missense Mutation ACA,ATA T162I NP_001374.3
Gene
OVCA2
Gene Name
ovarian tumor suppressor candidate 2
There are no transcripts associated with this gene.

View Full Product Details