Product Details

SNP ID
rs114111456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16235035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACCTATCCCTAACTTAACATCT[C/T]TTGTGTATATGAAACATTTTAAAGA
Phenotype
MIM: 605947
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIGL PubMed Links

Gene Details

Gene
PIGL
Gene Name
phosphatidylinositol glycan anchor biosynthesis class L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004278.3 Intron NP_004269.1
XM_011524080.2 Intron XP_011522382.1
XM_017025349.1 Intron XP_016880838.1
XM_017025350.1 Intron XP_016880839.1
XM_017025351.1 Intron XP_016880840.1
XM_017025352.1 Intron XP_016880841.1
XM_017025353.1 Intron XP_016880842.1
XM_017025354.1 Intron XP_016880843.1
XM_017025355.1 Intron XP_016880844.1
XM_017025356.1 Intron XP_016880845.1

View Full Product Details