Product Details

SNP ID
rs116025238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18970696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAATCGGAAACCTGGCTGAGAAC[A/C]ACTTGCCCAAGGCCGATGAGTGTCC
Phenotype
MIM: 615886
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM83G PubMed Links

Gene Details

Gene
FAM83G
Gene Name
family with sequence similarity 83 member G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039999.2 3386 Intron NP_001035088.2
XM_017024953.1 3386 UTR 3 XP_016880442.1
Gene
SLC5A10
Gene Name
solute carrier family 5 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042450.2 3386 Intron NP_001035915.1
NM_001270648.1 3386 Intron NP_001257577.1
NM_001270649.1 3386 Intron NP_001257578.1
NM_001282417.1 3386 Intron NP_001269346.1
NM_152351.4 3386 Intron NP_689564.3
XM_011523652.1 3386 Intron XP_011521954.1
XM_011523653.2 3386 Intron XP_011521955.1
XM_017024191.1 3386 Intron XP_016879680.1

View Full Product Details