Product Details

SNP ID
rs116004401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:760215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTATTGTGTTTGGCAAACCACTC[A/G]TCACTTTTATCTGCTGCCATTGCCT
Phenotype
MIM: 606969
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DBIL5P PubMed Links

Gene Details

Gene
DBIL5P
Gene Name
diazepam binding inhibitor-like 5, pseudogene
There are no transcripts associated with this gene.

Gene
GEMIN4
Gene Name
gem nuclear organelle associated protein 4
There are no transcripts associated with this gene.

Gene
GLOD4
Gene Name
glyoxalase domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016080.3 953 Silent Mutation GAC,GAT D285D NP_057164.3
XM_006721540.2 953 Silent Mutation GAC,GAT D276D XP_006721603.1
XM_017024711.1 953 Silent Mutation GAC,GAT D276D XP_016880200.1
XM_017024712.1 953 Silent Mutation GAC,GAT D229D XP_016880201.1

View Full Product Details