Product Details

SNP ID
rs114814106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:769915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGATCCATCTTAGAAAGTTCT[C/T]GAAATGCTTCATCCCCGACAAAGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLOD4 PubMed Links

Gene Details

Gene
GLOD4
Gene Name
glyoxalase domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016080.3 883 Missense Mutation CAA,CGA Q262R NP_057164.3
XM_006721540.2 883 Missense Mutation CAA,CGA Q253R XP_006721603.1
XM_017024711.1 883 Missense Mutation CAA,CGA Q253R XP_016880200.1
XM_017024712.1 883 Missense Mutation CAA,CGA Q206R XP_016880201.1

View Full Product Details