Product Details
- SNP ID
-
rs111661522
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:3609393 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTCGTGGTTGCCAGCGTGACTCTG[G/T]GCCAGGCCTGCCTGAGCCTGGGCTC
- Phenotype
-
MIM: 605060
MIM: 602076
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SHPK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3840876] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SHPK
- Gene Name
- sedoheptulokinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013276.2 |
19 |
UTR 3 |
|
|
NP_037408.2 |
- Gene
- TRPV1
- Gene Name
- transient receptor potential cation channel subfamily V member 1
View Full Product Details