Product Details

SNP ID
rs111972538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7289557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGGCTGCGGGCTGCCGGGGGCC[A/G]GGGGCCTGCTGGGGGCCAGGGGCCT
Phenotype
MIM: 138190 MIM: 611447
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC2A4 PubMed Links

Gene Details

Gene
SLC2A4
Gene Name
solute carrier family 2 member 4
There are no transcripts associated with this gene.

Gene
YBX2
Gene Name
Y-box binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015982.3 1074 Silent Mutation NP_057066.2
XM_017024713.1 1074 Intron XP_016880202.1

View Full Product Details