Product Details

SNP ID
rs115387034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7550764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGCTCTGGGACCCCCACTAGGG[C/T]CCGCTTTGCTCATCTGTCTTTCCTT
Phenotype
MIM: 602695 MIM: 604472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFSF12 PubMed Links

Gene Details

Gene
TNFSF12
Gene Name
tumor necrosis factor superfamily member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003809.2 Intron NP_003800.1
Gene
TNFSF12-TNFSF13
Gene Name
TNFSF12-TNFSF13 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172089.3 Intron NP_742086.1
Gene
TNFSF13
Gene Name
tumor necrosis factor superfamily member 13
There are no transcripts associated with this gene.

View Full Product Details