Product Details

SNP ID
rs113047719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7932536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCGAAGGGAAAGTACAAGGGAGC[G/T]GGAGCTGACCCTGGGTAGAACGGGT
Phenotype
MIM: 611425 MIM: 604111 MIM: 610969
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CNTROB PubMed Links

Gene Details

Gene
CNTROB
Gene Name
centrobin, centriole duplication and spindle assembly protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037144.5 288 Intron NP_001032221.1
NM_053051.3 288 Intron NP_444279.2
XM_005256437.2 288 UTR 5 XP_005256494.1
XM_005256438.2 288 UTR 5 XP_005256495.1
XM_005256439.2 288 UTR 5 XP_005256496.1
XM_017024128.1 288 UTR 5 XP_016879617.1
XM_017024129.1 288 Intron XP_016879618.1
XM_017024130.1 288 UTR 5 XP_016879619.1
XM_017024131.1 288 Intron XP_016879620.1
XM_017024132.1 288 UTR 5 XP_016879621.1
XM_017024133.1 288 Intron XP_016879622.1
XM_017024134.1 288 UTR 5 XP_016879623.1
XM_017024135.1 288 UTR 5 XP_016879624.1
XM_017024136.1 288 UTR 5 XP_016879625.1
XM_017024137.1 288 UTR 5 XP_016879626.1
XM_017024138.1 288 Intron XP_016879627.1
XM_017024139.1 288 UTR 5 XP_016879628.1
XM_017024140.1 288 UTR 5 XP_016879629.1
XM_017024141.1 288 UTR 5 XP_016879630.1
XM_017024142.1 288 UTR 5 XP_016879631.1
XM_017024143.1 288 Intron XP_016879632.1
XM_017024144.1 288 UTR 5 XP_016879633.1
Gene
KCNAB3
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 3
There are no transcripts associated with this gene.

Gene
TRAPPC1
Gene Name
trafficking protein particle complex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166621.1 288 Intron NP_001160093.1
NM_021210.4 288 Intron NP_067033.1

View Full Product Details