Product Details

SNP ID
rs117117111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8122239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGCCTCGGAGCAGAACCGGCCA[C/T]TCCCCAAGCCCACCATCCACCGCAG
Phenotype
MIM: 607206 MIM: 608059 MIM: 610407
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALOXE3 PubMed Links

Gene Details

Gene
ALOXE3
Gene Name
arachidonate lipoxygenase 3
There are no transcripts associated with this gene.

Gene
HES7
Gene Name
hes family bHLH transcription factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165967.1 Intron NP_001159439.1
NM_032580.3 Intron NP_115969.2
XM_011524041.2 Intron XP_011522343.1
XM_017025232.1 Intron XP_016880721.1
Gene
TRG-GCC2-6
Gene Name
transfer RNA-Gly (GCC) 2-6
There are no transcripts associated with this gene.

View Full Product Details