Product Details

SNP ID
rs115562095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:10444549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTAAATTCTATAAACTTTAGGCC[A/C]CAAAAAACCTGGATGTGCATCTGAA
Phenotype
MIM: 160742
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MYH4 PubMed Links

Gene Details

Gene
MYH4
Gene Name
myosin heavy chain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017533.2 Intron NP_060003.2
XM_017024676.1 Intron XP_016880165.1
Gene
MYHAS
Gene Name
myosin heavy chain gene cluster antisense RNA
There are no transcripts associated with this gene.

View Full Product Details