Product Details

SNP ID
rs117787091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37517531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGTCAAGCAGAAGCCTCAGCAA[C/T]AGCTGTTTAATGAAAATGTTCCACA
Phenotype
MIM: 606618 MIM: 607291
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP14 PubMed Links

Gene Details

Gene
DUSP14
Gene Name
dual specificity phosphatase 14
There are no transcripts associated with this gene.

Gene
SYNRG
Gene Name
synergin, gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163544.2 5561 UTR 3 NP_001157016.1
NM_001163545.2 5561 UTR 3 NP_001157017.1
NM_001163546.2 5561 UTR 3 NP_001157018.1
NM_001163547.2 5561 UTR 3 NP_001157019.1
NM_007247.5 5561 UTR 3 NP_009178.3
NM_080550.4 5561 UTR 3 NP_542117.3
NM_198882.2 5561 UTR 3 NP_942583.1
XM_005256980.4 5561 Intron XP_005257037.1
XM_017024076.1 5561 UTR 3 XP_016879565.1
XM_017024077.1 5561 UTR 3 XP_016879566.1
XM_017024078.1 5561 UTR 3 XP_016879567.1
XM_017024079.1 5561 UTR 3 XP_016879568.1
XM_017024080.1 5561 UTR 3 XP_016879569.1
XM_017024081.1 5561 UTR 3 XP_016879570.1
XM_017024082.1 5561 UTR 3 XP_016879571.1
XM_017024083.1 5561 UTR 3 XP_016879572.1
XM_017024084.1 5561 UTR 3 XP_016879573.1
XM_017024085.1 5561 UTR 3 XP_016879574.1
XM_017024086.1 5561 UTR 3 XP_016879575.1
XM_017024087.1 5561 UTR 3 XP_016879576.1
XM_017024088.1 5561 UTR 3 XP_016879577.1
XM_017024089.1 5561 UTR 3 XP_016879578.1
XM_017024090.1 5561 UTR 3 XP_016879579.1
XM_017024091.1 5561 UTR 3 XP_016879580.1
XM_017024092.1 5561 UTR 3 XP_016879581.1
XM_017024093.1 5561 UTR 3 XP_016879582.1
XM_017024094.1 5561 Intron XP_016879583.1
XM_017024095.1 5561 Intron XP_016879584.1
XM_017024096.1 5561 Intron XP_016879585.1
XM_017024097.1 5561 UTR 3 XP_016879586.1
XM_017024098.1 5561 Intron XP_016879587.1
XM_017024099.1 5561 UTR 3 XP_016879588.1
XM_017024100.1 5561 Intron XP_016879589.1
XM_017024101.1 5561 Intron XP_016879590.1
XM_017024102.1 5561 UTR 3 XP_016879591.1
XM_017024103.1 5561 Intron XP_016879592.1
XM_017024104.1 5561 Intron XP_016879593.1
XM_017024105.1 5561 Intron XP_016879594.1
XM_017024106.1 5561 UTR 3 XP_016879595.1

View Full Product Details