Product Details

SNP ID
rs112737273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50367884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGTGTGTCCCTAAATAGCATG[C/T]GTTCATGACGCTGGCTCACTTTATT
Phenotype
MIM: 610885 MIM: 611837 MIM: 608125
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EME1 PubMed Links

Gene Details

Gene
EME1
Gene Name
essential meiotic structure-specific endonuclease 1
There are no transcripts associated with this gene.

Gene
MRPL27
Gene Name
mitochondrial ribosomal protein L27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016504.2 686 UTR 3 NP_057588.1
Gene
XYLT2
Gene Name
xylosyltransferase 2
There are no transcripts associated with this gene.

View Full Product Details