Product Details

SNP ID
rs111565436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38674206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGCGGCGGTGTTAGGAACGCCG[A/G]GCCGCCCGGTCGGTGCCCAGAGGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf96 PubMed Links

Gene Details

Gene
C17orf96
Gene Name
chromosome 17 open reading frame 96
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130677.1 729 Missense Mutation CCC,CTC P97L NP_001124149.1

View Full Product Details