Product Details

SNP ID
rs118113570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:57839806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGGGAGGCAAGAGACCTTTCG[A/G]CTGGTCTGCAGGTGCCTCCAAATGC
Phenotype
MIM: 611985
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS23 PubMed Links

Gene Details

Gene
MRPS23
Gene Name
mitochondrial ribosomal protein S23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016070.3 612 Missense Mutation CCG,TCG P184S NP_057154.2

View Full Product Details