Product Details

SNP ID
rs113040772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58003164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGCACAAAAGCCTATGTTGTCT[A/T]TGACCACCAGAAAATAAGCAGCATC
Phenotype
MIM: 600812
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SRSF1 PubMed Links

Gene Details

Gene
SRSF1
Gene Name
serine and arginine rich splicing factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001078166.1 3398 UTR 3 NP_001071634.1
NM_006924.4 3398 UTR 3 NP_008855.1

View Full Product Details