Product Details

SNP ID
rs113668102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39202678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTATGCCAAGGATTCTGTACTT[C/T]CTAGTTTCAGAATGATCGAAGGAAG
Phenotype
MIM: 114207 MIM: 180466
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACNB1 PubMed Links

Gene Details

Gene
CACNB1
Gene Name
calcium voltage-gated channel auxiliary subunit beta 1
There are no transcripts associated with this gene.

Gene
RPL19
Gene Name
ribosomal protein L19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000981.3 Intron NP_000972.1
XM_005257564.3 Intron XP_005257621.1
Gene
STAC2
Gene Name
SH3 and cysteine rich domain 2
There are no transcripts associated with this gene.

View Full Product Details