Product Details

SNP ID
rs115348708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63820356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGCTCCACCTCCTTCTTACCAA[C/G]AGGCAACTGTCGTATCCGGTGCTGC
Phenotype
MIM: 613369 MIM: 601681
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DDX42 PubMed Links

Gene Details

Gene
DDX42
Gene Name
DEAD-box helicase 42
There are no transcripts associated with this gene.

Gene
FTSJ3
Gene Name
FtsJ homolog 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017647.3 2801 Missense Mutation CTT,GTT L719V NP_060117.3
Gene
PSMC5
Gene Name
proteasome 26S subunit, ATPase 5
There are no transcripts associated with this gene.

View Full Product Details