Product Details

SNP ID
rs116469710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63940783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGATGCTGCCTGCTAGACAAGAGA[C/T]TCCTTGACACCTGGGCGCACAGTCT
Phenotype
MIM: 147245 MIM: 603967
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD79B PubMed Links

Gene Details

Gene
CD79B
Gene Name
CD79b molecule
There are no transcripts associated with this gene.

Gene
SCN4A
Gene Name
sodium voltage-gated channel alpha subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000334.4 5576 Silent Mutation GAA,GAG E1833E NP_000325.4

View Full Product Details