Product Details

SNP ID
rs118115378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:69250533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCTCTTTGAAGGCGGTCTACTT[C/G]TAGGTTTTCTATCCAGTCCTTCAAT
Phenotype
MIM: 612508 MIM: 612503
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ABCA10 PubMed Links

Gene Details

Gene
ABCA10
Gene Name
ATP binding cassette subfamily A member 10
There are no transcripts associated with this gene.

Gene
ABCA5
Gene Name
ATP binding cassette subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018672.4 5842 Missense Mutation CAA,GAA Q1542E NP_061142.2
NM_172232.3 5842 Missense Mutation CAA,GAA Q1542E NP_758424.1

View Full Product Details