Product Details
- SNP ID
-
rs116036188
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:74208478 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAATTCAAGGAAAACCCTGGGTGC[A/G]TGTTGTGCGCAGTGGCTGGAGGCAT
- Phenotype
-
MIM: 604182
MIM: 608855
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MGC16275
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs571648436] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MGC16275
- Gene Name
- uncharacterized protein MGC16275
There are no transcripts associated with this gene.
- Gene
- RPL38
- Gene Name
- ribosomal protein L38
- Gene
- TTYH2
- Gene Name
- tweety family member 2
There are no transcripts associated with this gene.
View Full Product Details