Product Details

SNP ID
rs114678634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74611362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACACTGGCATCACTAGCTCAGG[A/T]TGAGCTCAGGCCTGGGGGCAGAGGG
Phenotype
MIM: 609801
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CD300E PubMed Links

Gene Details

Gene
CD300E
Gene Name
CD300e molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181449.2 2046 UTR 3 NP_852114.2
XM_017024575.1 2046 Intron XP_016880064.1
Gene
LOC101928343
Gene Name
uncharacterized LOC101928343
There are no transcripts associated with this gene.

View Full Product Details