Product Details
- SNP ID
-
rs115286715
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:78115976 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTACTCCTGGGGGCTGCCTCTCC[A/G]GTATGGGGTCCGACCCCCTTCTGCA
- Phenotype
-
MIM: 605828
MIM: 610741
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMC6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs572964531] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMC6
- Gene Name
- transmembrane channel like 6
- Gene
- TNRC6C
- Gene Name
- trinucleotide repeat containing 6C
There are no transcripts associated with this gene.
- Gene
- TNRC6C-AS1
- Gene Name
- TNRC6C antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details