Product Details

SNP ID
rs116980189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78796923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTATTTATTAAGTTACAAGTTGG[C/T]AGGCACAGCTTGAGCAACATAGAAA
Phenotype
MIM: 612543
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP36 PubMed Links

Gene Details

Gene
USP36
Gene Name
ubiquitin specific peptidase 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321291.1 4614 UTR 3 NP_001308220.1
XM_005257542.2 4614 UTR 3 XP_005257599.1
XM_005257543.2 4614 UTR 3 XP_005257600.1
XM_011525065.1 4614 UTR 3 XP_011523367.1
XM_011525066.2 4614 UTR 3 XP_011523368.1
XM_011525067.2 4614 Intron XP_011523369.1
XM_011525068.1 4614 UTR 3 XP_011523370.1
XM_011525069.2 4614 UTR 3 XP_011523371.1
XM_011525070.1 4614 UTR 3 XP_011523372.1
XM_011525071.2 4614 Intron XP_011523373.1
XM_017024900.1 4614 Intron XP_016880389.1
XM_017024901.1 4614 UTR 3 XP_016880390.1

View Full Product Details