Product Details

SNP ID
rs112619663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41177742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGTTAATAAAGGCAGGTGAGTA[C/T]AGGTGAGGGCATCAGCAGCAAGAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP4-1 PubMed Links

Gene Details

Gene
KRTAP4-1
Gene Name
keratin associated protein 4-1
There are no transcripts associated with this gene.

Gene
KRTAP4-2
Gene Name
keratin associated protein 4-2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033062.3 467 UTR 3 NP_149051.1
Gene
KRTAP4-3
Gene Name
keratin associated protein 4-3
There are no transcripts associated with this gene.

View Full Product Details