Product Details
- SNP ID
-
rs115695615
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41502347 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCACGGGTCCTGCAGTCACTATC[C/T]TAAGAAAGAGGCTGGAGAGGACCTA
- Phenotype
-
MIM: 148065
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT13
PubMed Links
Gene Details
- Gene
- KRT13
- Gene Name
- keratin 13
View Full Product Details