Product Details

SNP ID
rs111835360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41854193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGACAACATGATGCTGTCTGATA[A/G]AAGTTTGAATTGACCACAGACATTT
Phenotype
MIM: 608778
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL10 PubMed Links

Gene Details

Gene
KLHL10
Gene Name
kelch like family member 10
There are no transcripts associated with this gene.

Gene
KLHL11
Gene Name
kelch like family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018143.2 1735 Silent Mutation TTC,TTT F558F NP_060613.1

View Full Product Details