Product Details

SNP ID
rs112332573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42404933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGTAGACCGCGGTCTTGGAGCG[C/T]GCGTACACCACGTGGTCGGGCGTGA
Phenotype
MIM: 603198
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTRF PubMed Links

Gene Details

Gene
PTRF
Gene Name
polymerase I and transcript release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012232.5 1150 Silent Mutation GCA,GCG A309A NP_036364.2
XM_005257242.3 1150 Intron XP_005257299.1

View Full Product Details