Product Details

SNP ID
rs113459211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43015666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTGGCCCCATATCCTGAGCTGA[C/T]GTTTCAATTCTTTGGGGAGGTGGCG
Phenotype
MIM: 600735 MIM: 601555 MIM: 604631
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFI35 PubMed Links

Gene Details

Gene
IFI35
Gene Name
interferon induced protein 35
There are no transcripts associated with this gene.

Gene
RND2
Gene Name
Rho family GTPase 2
There are no transcripts associated with this gene.

Gene
VAT1
Gene Name
vesicle amine transport 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006373.3 1697 UTR 3 NP_006364.2

View Full Product Details