Product Details
- SNP ID
-
rs115182680
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:43848654 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACAGGGAAGAGCTGAGGGACCACC[A/G]GAAGTACTGGTGCAGGAAGGGTGGG
- Phenotype
-
MIM: 610520
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD300LG
PubMed Links
Gene Details
- Gene
- CD300LG
- Gene Name
- CD300 molecule like family member g
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001168322.1 |
580 |
Missense Mutation |
CAG,CGG |
Q47R |
NP_001161794.1 |
NM_001168323.1 |
580 |
Missense Mutation |
CAG,CGG |
Q47R |
NP_001161795.1 |
NM_001168324.1 |
580 |
Missense Mutation |
CAG,CGG |
Q47R |
NP_001161796.1 |
NM_145273.3 |
580 |
Missense Mutation |
CAG,CGG |
Q47R |
NP_660316.2 |
XM_005257075.2 |
580 |
Missense Mutation |
CAG,CGG |
Q47R |
XP_005257132.1 |
XM_005257076.2 |
580 |
Missense Mutation |
CAG,CGG |
Q47R |
XP_005257133.1 |
XM_006721721.2 |
580 |
Missense Mutation |
CAG,CGG |
Q47R |
XP_006721784.1 |
XM_006721722.2 |
580 |
Missense Mutation |
CAG,CGG |
Q47R |
XP_006721785.1 |
XM_011524384.2 |
580 |
Missense Mutation |
CAG,CGG |
Q47R |
XP_011522686.1 |
XM_017024235.1 |
580 |
Missense Mutation |
CAG,CGG |
Q47R |
XP_016879724.1 |
View Full Product Details