Product Details

SNP ID
rs112563021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29569324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTTTCCCCATGTATAGGTGTCA[C/G]CAAGAGTGACCTACACACGAGTGAG
Phenotype
MIM: 608434
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ABHD15 PubMed Links

Gene Details

Gene
ABHD15
Gene Name
abhydrolase domain containing 15
There are no transcripts associated with this gene.

Gene
GIT1
Gene Name
GIT ArfGAP 1
There are no transcripts associated with this gene.

Gene
TP53I13
Gene Name
tumor protein p53 inducible protein 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138349.2 186 Missense Mutation CCA,GCA P50A NP_612358.3
XM_017025282.1 186 Missense Mutation CCA,GCA P25A XP_016880771.1

View Full Product Details