Product Details

SNP ID
rs117301138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29571116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGCATCTAGCACAGCACTGAGC[A/G]CCAAGTCAACTCAATCAAAGAATAA
Phenotype
MIM: 608434
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD15 PubMed Links

Gene Details

Gene
ABHD15
Gene Name
abhydrolase domain containing 15
There are no transcripts associated with this gene.

Gene
GIT1
Gene Name
GIT ArfGAP 1
There are no transcripts associated with this gene.

Gene
TP53I13
Gene Name
tumor protein p53 inducible protein 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138349.2 Intron NP_612358.3
XM_017025282.1 Intron XP_016880771.1

View Full Product Details