Product Details

SNP ID
rs116750233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32932861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTATAGAAGGATATAATTTTGTC[C/T]TGAAGGGATATCCCTAACATCTTTG
Phenotype
MIM: 615949
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM98 PubMed Links

Gene Details

Gene
TMEM98
Gene Name
transmembrane protein 98
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033504.1 Intron NP_001028676.1
NM_001301746.1 Intron NP_001288675.1
NM_015544.2 Intron NP_056359.2

View Full Product Details