Product Details

SNP ID
rs114393730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35573676 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCGATGCGATGATGAGTGAAGT[A/G]GAGCCTGACCTGGTATTGCCATTGC
Phenotype
MIM: 601758
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105371743 PubMed Links

Gene Details

Gene
LOC105371743
Gene Name
uncharacterized LOC105371743
There are no transcripts associated with this gene.

Gene
PEX12
Gene Name
peroxisomal biogenesis factor 12
There are no transcripts associated with this gene.

Gene
SNORD7
Gene Name
small nucleolar RNA, C/D box 7
There are no transcripts associated with this gene.

View Full Product Details