Product Details

SNP ID
rs114139820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12094128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCGGGTACCGAGTCCGGCAGAA[A/G]GATCTGGGCATGATCCACAAAGCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD62 PubMed Links

Gene Details

Gene
ANKRD62
Gene Name
ankyrin repeat domain 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277333.1 333 Silent Mutation AAA,AAG K37K NP_001264262.1
XM_017025720.1 333 Silent Mutation AAA,AAG K37K XP_016881209.1

View Full Product Details