Product Details

SNP ID
rs116882881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12666503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCAGGCTGGAGTGCATTGTGCA[A/G]TCATGGCTCACTGAAGCCTCCTAAC
Phenotype
MIM: 609702 MIM: 609216
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP76 PubMed Links

Gene Details

Gene
CEP76
Gene Name
centrosomal protein 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271989.1 Intron NP_001258918.1
NM_024899.3 Intron NP_079175.2
XM_005258149.4 Intron XP_005258206.1
XM_017025981.1 Intron XP_016881470.1
XM_017025982.1 Intron XP_016881471.1
XM_017025983.1 Intron XP_016881472.1
XM_017025984.1 Intron XP_016881473.1
XM_017025985.1 Intron XP_016881474.1
XM_017025986.1 Intron XP_016881475.1
XM_017025987.1 Intron XP_016881476.1
Gene
PSMG2
Gene Name
proteasome assembly chaperone 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020232.4 Intron NP_064617.2
NM_147163.1 Intron NP_671692.1
Gene
SPIRE1
Gene Name
spire type actin nucleation factor 1
There are no transcripts associated with this gene.

View Full Product Details