Product Details

SNP ID
rs117709261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:13664173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCACATCTTCAGAGCATTTTTTA[C/G]TAGAGAATTTCATATCATTCAAGCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM210A PubMed Links

Gene Details

Gene
FAM210A
Gene Name
family with sequence similarity 210 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098801.1 3418 UTR 3 NP_001092271.1
NM_152352.3 3418 UTR 3 NP_689565.2

View Full Product Details