Product Details
- SNP ID
-
rs114739741
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:580528 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGTTCGGGAAGCATTTGACCTCTT[C/T]GACGTGGACGGAAGTGGGACCATCG
- Phenotype
-
MIM: 603187
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CETN1
PubMed Links
Gene Details
- Gene
- CETN1
- Gene Name
- centrin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004066.2 |
186 |
Silent Mutation |
TTC,TTT |
F40F |
NP_004057.1 |
View Full Product Details