Product Details

SNP ID
rs114097633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:22179186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAATGGGTTATAAATGATACCAA[C/T]TGAAGGGTGTGTCACTATCAATTAG
Phenotype
MIM: 601656
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GATA6 PubMed Links

Gene Details

Gene
GATA6
Gene Name
GATA binding protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005257.5 Intron NP_005248.2

View Full Product Details