Product Details

SNP ID
rs112874582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35369336 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACCAGTATGGGTTCGTCGATGC[C/T]GAATCAGAATTGCGCTTCGGCTGAA
Phenotype
MIM: 609600
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF396 PubMed Links

Gene Details

Gene
ZNF396
Gene Name
zinc finger protein 396
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322286.1 1019 Missense Mutation CAG,CGG Q296R NP_001309215.1
NM_001322290.1 1019 Missense Mutation CAG,CGG Q296R NP_001309219.1
NM_001322291.1 1019 Intron NP_001309220.1
NM_145756.2 1019 Missense Mutation CAG,CGG Q296R NP_665699.1
XM_006722432.3 1019 Missense Mutation CAG,CGG Q296R XP_006722495.1
XM_017025687.1 1019 Missense Mutation CAG,CGG Q296R XP_016881176.1
XM_017025688.1 1019 Missense Mutation CAG,CGG Q296R XP_016881177.1
XM_017025689.1 1019 Intron XP_016881178.1

View Full Product Details