Product Details

SNP ID
rs113831516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:23602394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTCCAGAGCATAAGAGACACCC[G/T]GGAAAGAAGAGCATTGGTGGCTAAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD29 PubMed Links

Gene Details

Gene
ANKRD29
Gene Name
ankyrin repeat domain 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308238.1 Intron NP_001295167.1
NM_173505.3 Intron NP_775776.2
XM_011525830.2 Intron XP_011524132.1
XM_011525831.2 Intron XP_011524133.1
XM_011525832.2 Intron XP_011524134.1
XM_017025561.1 Intron XP_016881050.1

View Full Product Details