Product Details

SNP ID
rs111897207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:54271367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTGCATATTGTGCAGGAGTTCA[C/T]GATCAAGTGTTGCCCACACCAAATG
Phenotype
MIM: 605252
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLI PubMed Links

Gene Details

Gene
POLI
Gene Name
DNA polymerase iota
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007195.2 483 Silent Mutation CAC,CAT H41H NP_009126.2
XM_005258188.3 483 UTR 5 XP_005258245.1
XM_005258192.4 483 Silent Mutation CAC,CAT H41H XP_005258249.1
XM_006722378.2 483 Silent Mutation CAC,CAT H41H XP_006722441.1
XM_011525796.2 483 UTR 5 XP_011524098.1
XM_011525797.1 483 Silent Mutation CAC,CAT H41H XP_011524099.1

View Full Product Details