Product Details

SNP ID
rs112130712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:20967783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTCATTCAGTTCCTTCTGATGT[C/T]TCACTACCATCTGGTTGAATTTCTC
Phenotype
MIM: 601702
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ROCK1 PubMed Links

Gene Details

Gene
ROCK1
Gene Name
Rho associated coiled-coil containing protein kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005406.2 4102 Missense Mutation AAA,AGA K1054R NP_005397.1
XM_011526137.2 4102 Missense Mutation AAA,AGA K591R XP_011524439.1

View Full Product Details