Product Details

SNP ID
rs116064131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37565232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAGATTAGAATTAGAAATAAAG[G/T]CTTTCCCACATTCTTTGCATTTATA
Phenotype
MIM: 613903
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF540 PubMed Links

Gene Details

Gene
ZNF540
Gene Name
zinc finger protein 540
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172225.2 1963 Intron NP_001165696.1
NM_001172226.2 1963 Intron NP_001165697.1
NM_152606.4 1963 Intron NP_689819.1
Gene
ZNF571
Gene Name
zinc finger protein 571
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290314.1 1963 Missense Mutation NP_001277243.1
NM_001321272.1 1963 Missense Mutation NP_001308201.1
NM_016536.4 1963 Missense Mutation NP_057620.3
XM_011527024.2 1963 Missense Mutation XP_011525326.1
XM_017026855.1 1963 Missense Mutation XP_016882344.1
XM_017026856.1 1963 Missense Mutation XP_016882345.1
XM_017026857.1 1963 Missense Mutation XP_016882346.1
XM_017026858.1 1963 Missense Mutation XP_016882347.1
XM_017026859.1 1963 Missense Mutation XP_016882348.1
XM_017026860.1 1963 Missense Mutation XP_016882349.1
Gene
ZNF571-AS1
Gene Name
ZNF571 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details