Product Details

SNP ID
rs112575357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38917533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACTGTTCGTGCTTCTGTTACAG[C/T]CCTGTGAGGGACGACCGGCACACGG
Phenotype
MIM: 604495 MIM: 612804
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCER2 PubMed Links

Gene Details

Gene
CCER2
Gene Name
coiled-coil glutamate rich protein 2
There are no transcripts associated with this gene.

Gene
NFKBIB
Gene Name
NFKB inhibitor beta
There are no transcripts associated with this gene.

Gene
SARS2
Gene Name
seryl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145901.1 Intron NP_001139373.1
NM_017827.3 Intron NP_060297.1

View Full Product Details