Product Details
- SNP ID
-
rs114133800
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39008438 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCTCTGTTTTTGCCTTTTTGTTA[A/C]AAAATCAGTATGTCTCATTCGGTCA
- Phenotype
-
MIM: 609099
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO27
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201539158] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FBXO27
- Gene Name
- F-box protein 27
View Full Product Details