Product Details

SNP ID
rs118188904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39604609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCAATACTCTGGCAGGTGCCAT[A/G]CAAACTGCCTGTGTCTTTGTCTGTT
Phenotype
MIM: 608717
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LGALS13 PubMed Links

Gene Details

Gene
LGALS13
Gene Name
galectin 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013268.2 63 Missense Mutation TAC,TGC Y8C NP_037400.1
XM_011526874.2 63 Intron XP_011525176.1
XM_017026715.1 63 Missense Mutation ATA,ATG I43M XP_016882204.1

View Full Product Details