Product Details

SNP ID
rs111409979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40691861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGAGGTGCAGGATCTAGGGCAC[A/G]AAGTTGGGAGTTCCCCAGCCCCAGG
Phenotype
MIM: 615567 MIM: 604018
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COQ8B PubMed Links

Gene Details

Gene
COQ8B
Gene Name
coenzyme Q8B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142555.2 2111 UTR 3 NP_001136027.1
NM_024876.3 2111 UTR 3 NP_079152.3
Gene
NUMBL
Gene Name
NUMB like, endocytic adaptor protein
There are no transcripts associated with this gene.

View Full Product Details